Cyclopentylacetic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Cyclopentylacetic acid

113907-1G
102892-972EA 43.7 USD
102892-972
Cyclopentylacetic acid ≥97%
Cyclopentylacetic acid

Hazard: Irritant

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
MDL Number: MFCD00001387
CAS Number: 1123-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR