2-Bromo-3-nitrobenzoyl chloride ≥95%

Supplier: Matrix Scientific

113899-1G
102898-232EA 932.31 USD
102898-232
2-Bromo-3-nitrobenzoyl chloride ≥95%
2-Bromo-3-nitrobenzoyl chloride

Hazard: Irritant

Formula: C₇H₃BrClNO₃
MDL Number: MFCD18392277
CAS Number: 6286-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR