2-((Difluoromethyl)thio)benzoyl chloride ≥97%

Supplier: Matrix Scientific

113888-1G
102892-956EA 400 USD
102892-956
2-((Difluoromethyl)thio)benzoyl chloride ≥97%
2-((Difluoromethyl)thio)benzoyl chloride

Hazard: Irritant

Formula: C₈H₅ClF₂OS
MW: 222.64 g/mol
MDL Number: MFCD01631519
CAS Number: 79676-60-1
UN: 3265
ADR: 8,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR