1-(4-Methylphenyl)cyclopentanecarbonyl chloride ≥97%

Supplier: Matrix Scientific

113881-1G
102892-942EA 738.46 USD
102892-942
1-(4-Methylphenyl)cyclopentanecarbonyl chloride ≥97%
1-(4-Methylphenyl)cyclopentanecarbonyl chloride

Hazard: Irritant

Formula: C₁₃H₁₅ClO
MDL Number: MFCD03424726
CAS Number: 82278-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR