2-Phenylacryloyl chloride ≥97%

Supplier: Matrix Scientific

113879-1G
102898-132EA 813.85 USD
102898-132
2-Phenylacryloyl chloride ≥97%
2-Phenylacryloyl chloride

Hazard: Irritant

Formula: C₉H₇ClO
MDL Number: MFCD25964035
CAS Number: 51491-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR