4-sulfamoylbenzoyl chloride ≥95%

Supplier: Matrix Scientific

113876-1G
102898-126EA 813.85 USD
102898-126
4-sulfamoylbenzoyl chloride ≥95%
4-sulfamoylbenzoyl chloride

Hazard: Irritant

Formula: C₇H₆ClNO₃S
MDL Number: MFCD09755960
CAS Number: 51594-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR