2,4,6-Trimethylphenylacetic chloride ≥95%

Supplier: Matrix Scientific

113873-1G
102898-120EA 690.77 USD
102898-120
2,4,6-Trimethylphenylacetic chloride ≥95%
2,4,6-Trimethylphenylacetic chloride

Hazard: Irritant

Formula: C₁₁H₁₃ClO
MDL Number: MFCD16879087
CAS Number: 25692-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR