2,6-Difluoro-4-methoxybenzoyl chloride ≥97%

Supplier: Matrix Scientific

113863-1G
102898-052EA 280 USD
102898-052
2,6-Difluoro-4-methoxybenzoyl chloride ≥97%
2,6-Difluoro-4-methoxybenzoyl chloride

Hazard: Irritant

Formula: C₈H₅ClF₂O₂
MDL Number: MFCD03094495
CAS Number: 125369-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR