2-(Difluoromethoxy)benzoyl chloride ≥97%

Supplier: Matrix Scientific

113862-1G
102898-050EA 958.46 USD
102898-050
2-(Difluoromethoxy)benzoyl chloride ≥97%
2-(Difluoromethoxy)benzoyl chloride

Hazard: Irritant

Formula: C₈H₅ClF₂O₂
MDL Number: MFCD03421646
CAS Number: 95155-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR