2-Chloro-4-(trifluoromethyl)benzoyl chloride ≥97%

Supplier: Matrix Scientific

113861-1G
102892-938EA 232.31 USD
102892-938
2-Chloro-4-(trifluoromethyl)benzoyl chloride ≥97%
2-Chloro-4-(trifluoromethyl)benzoyl chloride

Hazard: Irritant

Formula: C₈H₃Cl₂F₃O
MDL Number: MFCD07779332
CAS Number: 76286-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR