2-(Quinolin-2-yl)acetic acid ≥97%

Supplier: Matrix Scientific

113851-1G
102892-918EA 1443.08 USD
102892-918
2-(Quinolin-2-yl)acetic acid ≥97%
2-(Quinolin-2-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₉NO₂
MDL Number: MFCD00956755
CAS Number: 284477-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR