3-Ethoxy-2-thenoic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Ethoxythiophene-2-carboxylic acid
113835-1G
102892-884EA
183.08
USD
102892-884
3-Ethoxy-2-thenoic acid ≥97%
3-Ethoxy-2-thenoic acid
Hazard: Irritant
Formula:
C₇H₈O₃S MW: 172.2 g/mol |
MDL Number:
MFCD00205205 CAS Number: 139926-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...