1-(Phenylsulfonyl)-1H-pyrrole-2-carbonyl chloride ≥97%

Supplier: Matrix Scientific

113824-1G
102892-862EA 813.85 USD
102892-862
1-(Phenylsulfonyl)-1H-pyrrole-2-carbonyl chloride ≥97%
1-(Phenylsulfonyl)-1H-pyrrole-2-carbonyl chloride

Hazard: Irritant

Formula: C₁₁H₈ClNO₃S
MDL Number: MFCD26099144
CAS Number: 86688-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR