Pyridin-3-ylacetyl chloride hydrochloride ≥97%
Supplier: Matrix Scientific
113811-1G
102892-838EA
813.85
USD
102892-838
Pyridin-3-ylacetyl chloride hydrochloride ≥97%
Pyridin-3-ylacetyl chloride hydrochloride
Hazard: Irritant
Formula:
C₇H₇Cl₂NO |
MDL Number:
MFCD22575248 CAS Number: 120067-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...