2-Chloro-6-methylnicotinoyl chloride ≥97%

Supplier: Matrix Scientific

113810-1G
102892-836EA 329.23 USD
102892-836
2-Chloro-6-methylnicotinoyl chloride ≥97%
2-Chloro-6-methylnicotinoyl chloride

Hazard: Irritant

Formula: C₇H₅Cl₂NO
MDL Number: MFCD09953461
CAS Number: 39853-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR