4-Toluenesulfonylacetyl chloride ≥95%

Supplier: Matrix Scientific

113801-1G
102892-816EA 932.31 USD
102892-816
4-Toluenesulfonylacetyl chloride ≥95%
4-Toluenesulfonylacetyl chloride

Hazard: Irritant

Formula: C₉H₉ClO₃S
MDL Number: MFCD00087059
CAS Number: 99766-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR