2,3,3-Trichloroacryloyl chloride ≥95%

Supplier: Matrix Scientific

113797-1G
102892-808EA 958.46 USD
102892-808
2,3,3-Trichloroacryloyl chloride ≥95%
2,3,3-Trichloroacryloyl chloride

Hazard: Flammable/Corrosive/Light Sensitive/Refrigerate

Formula: C₃Cl₄O
MDL Number: MFCD00043968
CAS Number: 815-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR