1-Trifluoromethylcyclopropane-1-carbonyl chloride ≥97%

Supplier: Matrix Scientific

113787-1G
102892-786EA 1061.54 USD
102892-786
1-Trifluoromethylcyclopropane-1-carbonyl chloride ≥97%
1-Trifluoromethylcyclopropane-1-carbonyl chloride

Hazard: Irritant

Formula: C₅H₄ClF₃O
MDL Number: MFCD20484747
CAS Number: 848074-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR