4-Methoxycyclohexanecarbonyl chloride ≥95%

Supplier: Matrix Scientific

113786-1G
102892-784EA 958.46 USD
102892-784
4-Methoxycyclohexanecarbonyl chloride ≥95%
4-Methoxycyclohexanecarbonyl chloride

Hazard: Irritant

Formula: C₈H₁₃ClO₂
MDL Number: MFCD03424700
CAS Number: 123790-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR