1-[4-(Trifluoromethyl)phenyl]cyclopropanecarbonyl chloride ≥97%

Supplier: Matrix Scientific

113783-1G
102892-780EA 932.31 USD
102892-780
1-[4-(Trifluoromethyl)phenyl]cyclopropanecarbonyl chloride ≥97%
1-[4-(Trifluoromethyl)phenyl]cyclopropanecarbonyl chloride

Hazard: Irritant

Formula: C₁₁H₈ClF₃O
MDL Number: MFCD22381472
CAS Number: 960071-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR