4'-Methylbiphenyl-2-carbonyl chloride ≥97%

Supplier: Matrix Scientific

113781-1G
102892-776EA 884.62 USD
102892-776
4'-Methylbiphenyl-2-carbonyl chloride ≥97%
4'-Methylbiphenyl-2-carbonyl chloride

Hazard: Irritant

Formula: C₁₄H₁₁ClO
MDL Number: MFCD15523789
CAS Number: 114772-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR