5-Oxo-1-phenylpyrrolidine-3-carbonyl chloride ≥97%

Supplier: Matrix Scientific

113775-1G
102898-042EA 813.85 USD
102898-042
5-Oxo-1-phenylpyrrolidine-3-carbonyl chloride ≥97%
5-Oxo-1-phenylpyrrolidine-3-carbonyl chloride

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₂
MDL Number: MFCD11706931
CAS Number: 1071382-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR