3-Hydroxy-2-quinoxalinecarboxylic acid ≥97%

Supplier: Matrix Scientific

113770-1G
102898-070EA 932.31 USD
102898-070
3-Hydroxy-2-quinoxalinecarboxylic acid ≥97%
3-Hydroxy-2-quinoxalinecarboxylic acid

Hazard: Irritant

Formula: C₉H₅ClN₂O₂
CAS Number: 98591-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR