1H-Indole-5-carbonyl chloride ≥97%

Supplier: Matrix Scientific

113762-1G
102892-764EA 958.46 USD
102892-764
1H-Indole-5-carbonyl chloride ≥97%
1H-Indole-5-carbonyl chloride

Hazard: Corrosive

Formula: C₉H₆ClNO
MDL Number: MFCD11520769
CAS Number: 161397-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR