5-(Benzyloxy)-1H-indole-2-carbonyl chloride ≥97%

Supplier: Matrix Scientific

113759-1G
102892-758EA 813.85 USD
102892-758
5-(Benzyloxy)-1H-indole-2-carbonyl chloride ≥97%
5-(Benzyloxy)-1H-indole-2-carbonyl chloride

Hazard: Irritant

Formula: C₁₆H₁₂ClNO₂
MDL Number: MFCD28099703
CAS Number: 64892-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR