Tetrahydro-2H-pyran-2-carbonyl chloride ≥97%

Supplier: Matrix Scientific

113758-1G
102892-756EA 696.9 USD
102892-756
Tetrahydro-2H-pyran-2-carbonyl chloride ≥97%
Tetrahydro-2H-pyran-2-carbonyl chloride

Hazard: Irritant

Formula: C₆H₉ClO₂
CAS Number: 40053-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR