(±)-Mandelonitrile ≥97%

Supplier: Matrix Scientific

113752-10G 113752-50G
102892-742EA 15.38 USD
102892-742 102892-744
(±)-Mandelonitrile ≥97%
(±)-Mandelonitrile

Hazard: Irritant

Formula: C₈H₇NO
MW: 133.15 g/mol
MDL Number: MFCD00004487
CAS Number: 532-28-5
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR