3'-Chloro-o-formotoluidide ≥97%

Supplier: Matrix Scientific

Synonyms: N-(3-chloro-2-methylphenyl)formamide

113742-1G
102892-728EA 690.77 USD
102892-728
3'-Chloro-o-formotoluidide ≥97%
3'-Chloro-o-formotoluidide

Hazard: Irritant

Formula: C₈H₈ClNO
MW: 169.61 g/mol
MDL Number: MFCD00021009
CAS Number: 71862-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR