3-Fluoropropane-1-sulfonyl chloride ≥97%

Supplier: Matrix Scientific

113736-1G
102892-710EA 1298.46 USD
102892-710
3-Fluoropropane-1-sulfonyl chloride ≥97%
3-Fluoropropane-1-sulfonyl chloride

Hazard: Irritant

Formula: C₃H₆ClFO₂S
MDL Number: MFCD01697637
CAS Number: 461-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR