3-Chloro-5-fluorobenzene-1-sulfonyl chloride ≥95%

Supplier: Matrix Scientific

113732-1G
102892-702EA 958.46 USD
102892-702
3-Chloro-5-fluorobenzene-1-sulfonyl chloride ≥95%
3-Chloro-5-fluorobenzene-1-sulfonyl chloride

Hazard: Irritant

Formula: C₆H₃Cl₂FO₂S
MW: 229.06 g/mol
MDL Number: MFCD13185437
CAS Number: 1131397-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR