p-Toluenesulfinyl chloride ≥97%

Supplier: Matrix Scientific

113731-1G
102892-700EA 813.85 USD
102892-700
p-Toluenesulfinyl chloride ≥97%
p-Toluenesulfinyl chloride

Hazard: Irritant

Formula: C₇H₇ClOS
MDL Number: MFCD01630822
CAS Number: 10439-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR