5-(Bromomethyl)-1-methyl-1H-indazole ≥97%

Supplier: Matrix Scientific

113727-1G
102892-690EA 1175.38 USD
102892-690
5-(Bromomethyl)-1-methyl-1H-indazole ≥97%
5-(Bromomethyl)-1-methyl-1H-indazole

Hazard: Irritant

Formula: C₉H₉BrN₂
MDL Number: MFCD11052617
CAS Number: 1092961-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR