2-Phenoxybenzyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(Chloromethyl)-2-phenoxybenzene2-(Chloromethyl)diphenyl Ether

113726-1G
102892-688EA 447.69 USD
102892-688
2-Phenoxybenzyl chloride ≥97%
2-Phenoxybenzyl chloride

Hazard: Irritant

Formula: C₁₃H₁₁ClO
MW: 218.68 g/mol
MDL Number: MFCD00449749
CAS Number: 5888-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR