2-(Chloromethyl)biphenyl ≥97%

Supplier: Matrix Scientific

113725-1G
102892-686EA 958.46 USD
102892-686
2-(Chloromethyl)biphenyl ≥97%
2-(Chloromethyl)biphenyl

Hazard: Irritant

Formula: C₁₃H₁₁Cl
MDL Number: MFCD18072916
CAS Number: 38580-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR