2-(Chloromethyl)-5-methyl-2,3-dihydro-1,2-oxazol-3-one ≥97%

Supplier: Matrix Scientific

113720-1G
102892-676EA 813.85 USD
102892-676
2-(Chloromethyl)-5-methyl-2,3-dihydro-1,2-oxazol-3-one ≥97%
2-(Chloromethyl)-5-methyl-2,3-dihydro-1,2-oxazol-3-one

Hazard: Irritant

Formula: C₅H₆ClNO₂
MDL Number: MFCD19236557
CAS Number: 69513-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR