2-Cyclopentylacetamide ≥97%

Supplier: Matrix Scientific

113714-1G
102892-664EA 473.85 USD
102892-664
2-Cyclopentylacetamide ≥97%
2-Cyclopentylacetamide

Hazard: Irritant

Formula: C₇H₁₃NO
MDL Number: MFCD09863414
CAS Number: 933-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR