1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane ≥97%

Supplier: Matrix Scientific

Synonyms: 1,4,8,11-Tetramethylcyclam

113712-1G
102892-660EA 570.77 USD
102892-660
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane ≥97%
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane

Hazard: Irritant

Formula: C₁₄H₃₂N₄
MW: 256.44 g/mol
MDL Number: MFCD00005106
CAS Number: 41203-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR