Glycidyl isobutyl ether ≥97%

Supplier: Matrix Scientific

113704-1G
102897-998EA 121.54 USD
102897-998
Glycidyl isobutyl ether ≥97%
Glycidyl isobutyl ether

Hazard: Irritant

Formula: C₇H₁₄O₂
MDL Number: MFCD00055638
CAS Number: 3814-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR