1-Benzyl-3-pyrroline ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Benzyl-2,5-dihydropyrrole

113693-1G 113693-10G
102892-630EA 727.69 USD
102892-630 102892-628
1-Benzyl-3-pyrroline ≥97%
1-Benzyl-3-pyrroline

Hazard: Corrosive/Air Sensitive

Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 60 °C (1 mmHg)
Density: 0.837
MDL Number: MFCD00012324
CAS Number: 6913-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR