N-(2-Hydroxyethyl)succinimide ≥97%

Supplier: Matrix Scientific

113692-1G
102892-626EA 73.85 USD
102892-626
N-(2-Hydroxyethyl)succinimide ≥97%
N-(2-Hydroxyethyl)succinimide

Hazard: Irritant

Formula: C₆H₉NO₃
MDL Number: MFCD00078332
CAS Number: 18190-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR