Isothiazolin-3-one ≥97%

Supplier: Matrix Scientific

Synonyms: Isothiazol-3(2H)-one

113690-1G
102892-622EA 298.46 USD
102892-622
Isothiazolin-3-one ≥97%
Isothiazolin-3-one

Hazard: Irritant

Formula: C₃H₃NOS
MW: 101.13 g/mol
MDL Number: MFCD09834764
CAS Number: 1003-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR