Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-Amino-4',5'-methylenedioxyacetophenone ≥97%

Supplier: Matrix Scientific


113688-1G
102892-618EA 92.85 USD InStock
102892-618
2'-Amino-4',5'-methylenedioxyacetophenone ≥97%
2'-Amino-4',5'-methylenedioxyacetophenone
Purity: 97%
CAS: 28657-75-2
MDL Number: MFCD00075855
Molecular Formula: C9H9NO3
Molecular Weight: 179,18
Hazard: Irritant


Formula: C₉H₉NO₃
MDL Number: MFCD00075855
CAS Number: 28657-75-2
EINECS: 249-131-7
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty