Benzyl-3-oxo-4-(prop-2-ynyl)piperazine-1-carboxylate ≥97%

Supplier: Matrix Scientific

113685-1G
102892-612EA 507.69 USD
102892-612
Benzyl-3-oxo-4-(prop-2-ynyl)piperazine-1-carboxylate ≥97%
Benzyl-3-oxo-4-(prop-2-ynyl)piperazine-1-carboxylate

Hazard: Irritant/Store in Freezer

Formula: C₁₅H₁₆N₂O₃
MW: 272.31 g/mol
MDL Number: MFCD12911962
CAS Number: 234108-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR