(4-Hydroxypiperidin-1-yl)piperidin-3-ylmethanone ≥97%

Supplier: Matrix Scientific

113682-1G
102892-606EA 932.31 USD
102892-606
(4-Hydroxypiperidin-1-yl)piperidin-3-ylmethanone ≥97%
(4-Hydroxypiperidin-1-yl)piperidin-3-ylmethanone

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD03425213
CAS Number: 496057-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR