4-(2-Methoxyphenyl)piperidine ≥97%

Supplier: Matrix Scientific

113681-1G
102892-602EA 150.77 USD
102892-602
4-(2-Methoxyphenyl)piperidine ≥97%
4-(2-Methoxyphenyl)piperidine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD00277219
CAS Number: 58333-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR