DL-3-Camphorcarboxylic acid ≥97%

Supplier: Matrix Scientific

113669-1G
102892-566EA 473.85 USD
102892-566
DL-3-Camphorcarboxylic acid ≥97%
DL-3-Camphorcarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₆O₃
MDL Number: MFCD08272794
CAS Number: 474-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR