3-Nitro-4-fluorobenzoyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-3-nitrobenzoyl chloride

113667-1G
102892-562EA 253.85 USD
102892-562
3-Nitro-4-fluorobenzoyl chloride ≥97%
3-Nitro-4-fluorobenzoyl chloride

Hazard: Irritant

Formula: C₇H₃ClFNO₃
MW: 203.56 g/mol
MDL Number: MFCD07368701
CAS Number: 400-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR