4-(Methylthio)phenylacetyl chloride ≥97%

Supplier: Matrix Scientific

113662-1G
102892-554EA 958.46 USD
102892-554
4-(Methylthio)phenylacetyl chloride ≥97%
4-(Methylthio)phenylacetyl chloride

Hazard: Irritant

Formula: C₉H₉ClOS
MDL Number: MFCD22575246
CAS Number: 63327-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR