2-Cyclohexylideneacetic acid ≥97%

Supplier: Matrix Scientific

113645-1G
102897-966EA 690.77 USD
102897-966
2-Cyclohexylideneacetic acid ≥97%
2-Cyclohexylideneacetic acid

Hazard: Irritant

Formula: C₈H₁₂O₂
MW: 140.18 g/mol
MDL Number: MFCD01815140
CAS Number: 1552-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR