2-Piperazin-1-ylnicotinic acid ≥97%

Supplier: Matrix Scientific

113640-1G
102897-956EA 958.46 USD
102897-956
2-Piperazin-1-ylnicotinic acid ≥97%
2-Piperazin-1-ylnicotinic acid

Hazard: Irritant

Formula: C₁₀H₁₃N₃O₂
MDL Number: MFCD05182223
CAS Number: 374063-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR